Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Disposal Cabinet
Disposal Cabinet
Connecting frame for disposal cabinet/dispenser unit, 80×30×160 mm
Catalog # 134-0487
Supplier:   Palbam
Disposal Cabinet
Catalog # 134-0487
Supplier:  Palbam
Disposal Cabinet
Disposal Cabinet
Connecting frame for disposal cabinet/dispenser unit, 80×30×160 mm
Price
£652.00
Log in for availability
1 * 1 items
Product Details & Documents
This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
  • Made of stainless steel 304 with electropolished finish
  • Two waste openings, two doors and levelling feet
  • Does not require connecting to the wall
Can be used in cleanroom and critical environments.
Certifications: ISO certified.
Delivery information: Typically available from stock for direct shipment from the supplier. Supplied with two large plastic bins.
Specifications
  • Description:
    Connecting frame for disposal cabinet/dispenser unit
  • Seq:
    02
  • W×D×H:
    80×30×160 mm