Order Entry
United Kingdom
ContactUsLinkComponent
Flask volumetric, capacity 1l, with rim, DURAN, class A, tolerance 0.4ml, height 300mm, graduation color blue, conformity certified. 1 * 1 items
Catalog # HIRS2800193_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 1l, with rim, DURAN, class A, tolerance 0.4ml, height 300mm, graduation color blue, conformity certified. 1 * 1 items
Catalog # HIRS2800193_S
Supplier:  HIRSCHMANN
Frequently Bought Together