Order Entry
United Kingdom
ContactUsLinkComponent
Flask volumetric, capacity 25ml, with rim, DURAN, class A, tolerance 0.04ml, height 110mm, graduation color blue, conformity certifi ed. 1 * 1 items
Catalog # HIRS2800170_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 25ml, with rim, DURAN, class A, tolerance 0.04ml, height 110mm, graduation color blue, conformity certifi ed. 1 * 1 items
Catalog # HIRS2800170_S
Supplier:  HIRSCHMANN
Frequently Bought Together