Order Entry
United Kingdom
ContactUsLinkComponent
Flask volumetric, capacity 2l, class A, amber graduated, glass stoppered, tolerance 0.6ml, height 370mm, NS-ground 29/32, graduation color white, conformity certified. 1 * 1 items
Catalog # HIRS2640294_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 2l, class A, amber graduated, glass stoppered, tolerance 0.6ml, height 370mm, NS-ground 29/32, graduation color white, conformity certified. 1 * 1 items
Catalog # HIRS2640294_S
Supplier:  HIRSCHMANN
Frequently Bought Together