Order Entry
United Kingdom
ContactUsLinkComponent
Pipette volumetric, capacity 10ml, class AS, amber graduation, soda-lime glass, tolerance 0.02ml, total length +-10mm:440mm, conform ity certified. 1 * 1 items
Catalog # HIRS1340160_S
Supplier:  HIRSCHMANN
undefined
Pipette volumetric, capacity 10ml, class AS, amber graduation, soda-lime glass, tolerance 0.02ml, total length +-10mm:440mm, conform ity certified. 1 * 1 items
Catalog # HIRS1340160_S
Supplier:  HIRSCHMANN
Frequently Bought Together