Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Brij® 35

Brij® 35

Glass bottle for solids, 250 g
Catalog # A15809.30
Irritant
Brij® 35
Catalog # A15809.30
CAS Number:  9002-92-0
Brij® 35
Brij® 35

Glass bottle for solids, 250 g

CAS Number:  9002-92-0
Price
£21.10
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 9002-92-0 Molecular Formula: C58H118O24 Formula Weight: 1199.57 Physical Form: Waxy solid Appearance: White Melting Point: ca 35-40° MDL No.: MFCD00080891

For use in Stein-Moore chromatography

Synonyms:  Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
  • CAS Number:9002-92-0
  • Formula:C₅₈H₁₁₈O₂₄
  • MDL Number:MFCD00080891
  • Molecular Weight:1199.56 g/mol
  • Melting Pt:∼ 35…40 °C
  • Storage Temperature:Ambient
  • EINECS:500-002-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.