Sample loops
Sample loop, PEEK, 20 µl
Catalog # VARIVLCZSL20PK
Jump to:
Discontinued
Product Details & Documents
The sample loops is used for valco C2 peak valve.
Specifications
- Capacity:20 µl
- Description:Sample loop
- Material:PEEK
- Seq:01