Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cyclohexanecarboxylic acid ≥98%

Cyclohexanecarboxylic acid ≥98%

Glass bottle for solids, 250 g
Catalog # A14693.30
Irritant
Cyclohexanecarboxylic acid ≥98%
Catalog # A14693.30
CAS Number:  98-89-5
Cyclohexanecarboxylic acid ≥98%
Cyclohexanecarboxylic acid ≥98%

Glass bottle for solids, 250 g

CAS Number:  98-89-5
Price
£32.60
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 98-89-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 118°(244°F) Refractive Index: 1.4610 Melting Point: 29-32° Boiling Point: 231-233° Beilstein Reference: 970529 Merck Reference: 14,2724 MDL No.: MFCD00001461

Synonyms:  Hexahydrobenzoic acid
  • CAS Number:98-89-5
  • Formula:C₇H₁₂O₂
  • MDL Number:MFCD00001461
  • Molecular Weight:128.17 g/mol
  • Merck Index:14,2724
  • Boiling Pt:231…233 °C
  • Melting Pt:29…32 °C
  • Density:1.048
  • Flash Pt:118 °C (244 °F)
  • Storage Temperature:Ambient
  • EINECS:202-711-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.