Objectives for CKX53
Plan achromatic objective, 10,5 mm
Catalog # OLYMN1215800
Supplier: OLYMPUS OPTICAL
Jump to:
Product Details & Documents
Specifications
- Magnification:10×/0,25
- Description:Plan achromatic objective
- Working distance:10,5 mm