Enterolactone ELISA kit
A competitive assay for measurement of enterolactone in plasma, urine, and other biological sample matrices.
- Measure enterolactone in plasma, urine, and other sample matrices
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure enterolactone levels down to 70 pg/ml
- Colorimetric reading at 405 to 420 nm
Enterolactone is a mammalian lignan with an estrogen-like diphenolic structure. It is produced by intestinal bacteria from two plant precursors (matairesinol and secoisolariciresinol) obtained in the diet. Enterolactone and other lignans and phytoestrogens have been associated with a reduced risk of acute coronary events, hormone-dependent cancers, and possibly osteoporosis. Several studies have suggested that serum enterolactone may serve as a biomarker of a healthy, high-fiber diet. Cayman Chemical’s Enterolactone ELISA kit is a competitive assay that utilizes a standard curve ranging from 15,6 to 2000 pg/ml. The assay has a range from 15,6 to 2500 pg/ml and a sensitivity (80% B/B0) of approximately 70 pg/ml.
: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate; polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.
- Pk:480 Tests
- Description:Enterolactone ELISA kit, 480 strip wells
- Cat. No.:CAYM500520-480
- Developmental time:90 - 120 min
- Sensitivity:70 pg/ml
- Detection range:15,6 - 2500 pg/ml
- Incubation time:18 hr
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



