Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Septa for 8 mm screw thread vials

Septa for 8 mm screw thread vials

Red PTFE/silicone, 8 mm, 1,52 mm
Catalog # REST21179
Supplier:   Restek
Septa for 8 mm screw thread vials
Catalog # REST21179
Supplier:  Restek
Septa for 8 mm screw thread vials
Septa for 8 mm screw thread vials

Red PTFE/silicone, 8 mm, 1,52 mm

Price
£324.00
Log in for availability
1 * 1.000 items
Product Details & Documents
Specifications
  • Ø:
    8 mm
  • Type:
    Red PTFE/silicone
  • Seq:
    02
  • Thickness:
    1,52 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.