Congo red
5 g
Catalog # ROTH0322.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), CI 22120, Direct Red 28
- CAS Number:573-58-0
- Formula:C₃₂H₂₂N₆Na₂O₆S₂
- MDL Number:MFCD00004028
- Molecular Weight:696.67 g/mol
- Merck Index:12,02562
- Melting Pt:> 360 °C
- Storage Temperature:Ambient
- EINECS:209-358-4
Specifications
Specifications
- Pk:5 g



