Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Ethanol absolute

Glass bottle, 1 L
Catalog # ROTH9065.1
Flammable
Marketsource
Supplier:   Roth Carl
Ethanol absolute
Catalog # ROTH9065.1
Supplier:  Roth Carl
Marketsource
CAS Number:  64-17-5
Ethanol absolute

Glass bottle, 1 L

CAS Number:  64-17-5
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Merck Index:12,03806
  • Boiling Pt:78.3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0.7895 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.