Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Propanol

2-Propanol

Glass bottle, 1 L
Catalog # ROTH6752.1
Flammable
Irritant
Marketsource
   Safer Human and Environmental Health
Supplier:   Roth Carl
2-Propanol
Catalog # ROTH6752.1
Supplier:  Roth Carl
Marketsource
   Safer Human and Environmental Health
CAS Number:  67-63-0
2-Propanol
2-Propanol

Glass bottle, 1 L

CAS Number:  67-63-0
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    ROTH6752.1
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications