Silver(I) oxide
10 g
Catalog # ROTH5309.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: di-Silver oxide, Disilver oxide, Silver oxide
- CAS Number:20667-12-3
- Formula:Ag₂O
- MDL Number:MFCD00003404
- Molecular Weight:231.74 g/mol
- Merck Index:12,08665
- Melting Pt:800…830 °C
- Density:7.14 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:243-957-1
- UN:1479
- ADR:5.1,II
Specifications
Specifications
- Pk:10 g



