2-Phenoxyethanol
Glass bottle, 1 L
Catalog # ROTH4348.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Merck Index:12,07410
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:13 °C
- Density:1.1094 g/cm³ (20 °C)
- Flash Pt:121 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:204-589-7
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



