Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Phenoxyethanol

2-Phenoxyethanol

Glass bottle, 250 mL
Catalog # ROTH4348.1
Irritant
Marketsource
Supplier:   Roth Carl
2-Phenoxyethanol
Catalog # ROTH4348.1
Supplier:  Roth Carl
Marketsource
CAS Number:  122-99-6
2-Phenoxyethanol
2-Phenoxyethanol

Glass bottle, 250 mL

CAS Number:  122-99-6
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Merck Index:12,07410
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:13 °C
  • Density:1.1094 g/cm³ (20 °C)
  • Flash Pt:121 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:204-589-7
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.