Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Palmitic acid

Glass bottle for solids, 1 kg
Catalog # ROTH5907.1
Marketsource
Supplier:   Roth Carl
Palmitic acid
Catalog # ROTH5907.1
Supplier:  Roth Carl
Marketsource
CAS Number:  57-10-3
Palmitic acid

Glass bottle for solids, 1 kg

CAS Number:  57-10-3
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256.43 g/mol
  • Merck Index:13,07064
  • Boiling Pt:352 °C (1013 hPa)
  • Melting Pt:61…63 °C
  • Density:0.853 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-312-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.