Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, micro

VWR®, Magnetic stirring bars, micro

13 mm, 3 mm
Catalog # 442-0470
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, micro
Catalog # 442-0470
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, micro
VWR®, Magnetic stirring bars, micro

13 mm, 3 mm

Price
£14.20
Log in for availability
1 * 10 items
Product Details & Documents

PTFE coated, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.

  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
  • Ø:
    3 mm
  • Length:
    13 mm
  • Seq:
    79
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.