Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

VWR®, Magnetic stirring bars, micro

8 mm, 1,5 mm
Catalog # 442-0462
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, micro
Catalog # 442-0462
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, micro
VWR®, Magnetic stirring bars, micro

8 mm, 1,5 mm

Price
£12.50
Log in for availability
1 * 10 items
Product Details & Documents

PTFE coated, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.

  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
  • Ø:
    1,5 mm
  • Length:
    8 mm
  • Seq:
    36