Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, micro

VWR®, Magnetic stirring bars, micro

2 mm, 2 mm
Catalog # 442-0449
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, micro
Catalog # 442-0449
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, micro
VWR®, Magnetic stirring bars, micro

2 mm, 2 mm

Price
£15.80
Log in for availability
1 * 10 items
Product Details & Documents

PTFE coated, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.

  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
  • Ø:
    2 mm
  • Length:
    2 mm
  • Seq:
    71