Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring

VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring

22 mm, 8 mm
Catalog # 442-0432
Supplier:   VWR Collection
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
Catalog # 442-0432
Supplier:  VWR Collection
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring

22 mm, 8 mm

Price
£22.00
Log in for availability
1 * 10 items
Product Details & Documents

PTFE covered, strong Alnico V magnetic core. Smooth surface. For use in containers with bases that are slightly uneven or curved.

  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)

Caution: Coloured PTFE coatings are not as inert as pure PTFE.

Specifications
  • Ø:
    8 mm
  • Length:
    22 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.