Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
13 mm, 8 mm
Catalog # 442-0428
Supplier:   VWR Collection
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
Catalog # 442-0428
Supplier:  VWR Collection
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
13 mm, 8 mm
Price
£12.50
Log in for availability
1 * 10 items
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. For use in containers with bases that are slightly uneven or curved.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
 : Coloured PTFE coatings are not as inert as pure PTFE.
Specifications
  • Ø:
    8 mm
  • Length:
    13 mm