Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, micro

VWR®, Magnetic stirring bars, micro

6 mm, 3 mm
Catalog # 442-0362
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, micro
Catalog # 442-0362
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, micro
VWR®, Magnetic stirring bars, micro

6 mm, 3 mm

Price
£30.60
Log in for availability
1 * 10 items
Product Details & Documents

PTFE coated, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.

  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
  • Ø:
    3 mm
  • Length:
    6 mm
  • Seq:
    04