Phenethyl bromide
Glass bottle for solids, 250 g
Catalog # FLUO019261-250G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
Synonyms: (2-Bromoethyl)benzene, 2-Phenylethyl bromide
- CAS Number:103-63-9
- Formula:BrCH₂CH₂C₆H₅
- MDL Number:MFCD00000240
- Molecular Weight:185.06 g/mol
- Boiling Pt:221 °C (1013 hPa)
- Melting Pt:–56 °C
- Density:1,37 g/cm³ (20 °C)
- Flash Pt:89 °C
- Storage Temperature:Refrigerator
- EINECS:203-130-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



