Disintegration testing accessories, QLA
Disintegration beaker with flat top, clear glass, 900 ml
QLAAGLA905-DB
Disintegration testing accessories and glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
: Complies to USP.
- Description:Disintegration beaker with flat top, clear glass, 900 ml
- Seq:08