Disintegration testing accessories, QLA
Disintegration beaker with flared top, clear glass, 900 ml
Catalog # QLAAGLA900-DB
Supplier: QUALITY LAB ACCESSORIES
Jump to:
Product Details & Documents
Disintegration testing accessories and glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
: Complies to USP.
Specifications
- Description:Disintegration beaker with flared top, clear glass, 900 ml