Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Dissolution calibration tools
Basket height spacer for legacy Vankel baths, 25 mm
  QLAADPSPBK-VK
Dissolution calibration tools
  QLAADPSPBK-VK
Dissolution calibration tools
Dissolution calibration tools
Basket height spacer for legacy Vankel baths, 25 mm
£105.00
1 * 1 items
These dissolution calibration tools are suitable for most dissolution baths.
  • Mechanical calibration parameters can easily be verified by using calibration tools
  • All calibration tools are serialised
 : Supplied with NIST traceable calibration certificates and user manuals.
  • Description:
    Basket height spacer for legacy Vankel baths, 25 mm