Disintegration testing accessories, QLA
Glass replacement tubes for 6 tube disintegration assembly, 25 mm
Catalog # QLAADISTUB-06
Supplier: QUALITY LAB ACCESSORIES
Jump to:
Product Details & Documents
Disintegration testing accessories and glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
Certifications: Complies to USP.
Specifications
- Description:Glass replacement tubes for 6 tube disintegration assembly, 25 mm
- Seq:03