Dissolution calibration tools
Depth set gauge, stainless steel, Vankel style, 25 mm
Catalog # QLAADEPGAG-VK
Supplier: QUALITY LAB ACCESSORIES
Jump to:
Product Details & Documents
These dissolution calibration tools are suitable for most dissolution baths.
- Mechanical calibration parameters can easily be verified by using calibration tools
- All calibration tools are serialised
Delivery information: Supplied with NIST traceable calibration certificates and user manuals.
Specifications
- Description:Depth set gauge, stainless steel, Vankel style, 25 mm