Dissolution vessel covers
Low evaporation conical cover, clear, for Hanson SR8-Plus Baths
QLAACOVERH-LL
Standard and low loss vessel covers for most dissolution baths.
- Slots and holes for temperature verification, sample introduction and sample collection should be closed when not in use
- Condensation is reduced by using vessel covers with conical bottoms that allow condensation to return back to vessel
The loss of media due to evaporation can be considerable. Vesssel covers reduce these issues and provide openings for cannula, cannula stoppers, temperature verification, and sample introduction. There are many varieties of covers to choose from, including one-piece, hinged, conical and rotary style.
: All QLA vessel covers are USP compliant and that they meet machine manufacturer specifications.
- Description:Low evaporation conical cover, clear, for Hanson SR8-Plus Baths