Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Dissolution baskets, QLA
Suppository basket, plastic
  QLAABSKSUP-CA
Dissolution baskets, QLA
  QLAABSKSUP-CA
Dissolution baskets, QLA
Dissolution baskets, QLA
Suppository basket, plastic
£373.00
1 * 1 items
These are clip style and O-ring push-on style baskets for most dissolution baths. All baskets are laser marked with a unique serial number (except suppository baskets).
  • 40 mesh baskets are made from sintered mesh
  • Different mesh sizes are available
The sintering process essentially creates a welded joint at each overlapping wire junction. Sintered baskets are stronger and last longer. They retain original dimensions and geometry and resist wobble. Most conventional baskets are not sintered and weaken over time. Critical basket geometry is verified on an optical comparator.
 : All baskets are supplied with a certificate of conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications.
 : Certificate of analysis (CoA) has to be ordered separately.
  • Description:
    Suppository basket, plastic
Accessories
  •  
    Description
  • QLAABSKHLD-12
    Basket holder, 12 position, universal, white acetal plastic
    1 * 1 items - £342.00
  •  
    Description
  • QLAABSKTOOL
    Basket insertion and removal tool for clip style baskets, black acetal plastic
    1 * 1 items - £357.11