Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Dissolution baskets, QLA
Dissolution baskets, QLA
40 Mesh basket, standard push-on style, 316 stainless steel, serialised
Catalog # QLAABSK040-DK
Dissolution baskets, QLA
Catalog # QLAABSK040-DK
Dissolution baskets, QLA
Dissolution baskets, QLA
40 Mesh basket, standard push-on style, 316 stainless steel, serialised
Price
£225.00
Log in for availability
1 * 1 items
Product Details & Documents
These are clip style and O-ring push-on style baskets for most dissolution baths. All baskets are laser marked with a unique serial number (except suppository baskets).
  • 40 mesh baskets are made from sintered mesh
  • Different mesh sizes are available
The sintering process essentially creates a welded joint at each overlapping wire junction. Sintered baskets are stronger and last longer. They retain original dimensions and geometry and resist wobble. Most conventional baskets are not sintered and weaken over time. Critical basket geometry is verified on an optical comparator.
Certifications: All baskets are supplied with a certificate of conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications.
Ordering information: Certificate of analysis (CoA) has to be ordered separately.
Specifications
  • Description:
    40 Mesh basket, standard push-on style, 316 stainless steel, serialised
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Basket holder, 12 position, universal, white acetal plastic
    Basket holder, 12 position, universal, white acetal plastic
    QLAABSKHLD-12
    Log in for availability
    1 * 1 items - £342.00
Product Information
  • Catalog #
    Availability
    Price
  • Basket insertion and removal tool for clip style baskets, black acetal plastic
    Basket insertion and removal tool for clip style baskets, black acetal plastic
    QLAABSKTOOL
    Log in for availability
    1 * 1 items - £357.11