Accessories for low temperature viscometer baths, mini
Bath oil, silicone, 2 l, for filling viscometer baths, with the temperature range of 100 - 150 °C
Catalog # STAN94631-0
Supplier: STANHOPE SETA
Accessories for low temperature viscometer baths, mini
Catalog # STAN94631-0
Supplier: STANHOPE SETA
Jump to:
Product Details & Documents
Specifications
- Description:Bath oil, silicone, 2 l, for filling viscometer baths, with the temperature range of 100 - 150 °C
- Seq:06