Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Büchner funnels, Hartley pattern
Spare base, 125 mm
  511-0842
Büchner funnels, Hartley pattern
  511-0842
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
Spare base, 125 mm
£318.00
1 * 1 items
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
  • Ø:
    125 mm
  • Description:
    Spare base