Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Büchner funnels, Hartley pattern
Spare base, 70 mm
Catalog # 511-0840
Supplier:   DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Catalog # 511-0840
Supplier:  DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
Spare base, 70 mm
Price
£147.00
Log in for availability
1 * 1 items
Product Details & Documents
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
Specifications
  • Ø:
    70 mm
  • Description:
    Spare base