Büchner funnels, Hartley pattern
90 mm
511-0828
Top and base: Borosilicate glass with support plate.
- Available with choice of perforated disc material
- Complete with three PE coated clips
- Available for 4 filter sizes
Three piece construction.
- For filter Ø:90 mm