Gas sample tubes
200 ml, with two 1-way capillary stopcocks
Catalog # 201-0453
Supplier: DIXON SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass, grade 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Two capillary one-way stopcocks or two capillary three-way stopcocks with threaded stopcock arrester.
Specifications
- Type:with two 1-way capillary stopcocks
- Capacity:200 ml