Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Test tubes, reductase
Test tubes, reductase
Tubes with marks at 5 and 10 ml for Phosphatase and COD tests
Catalog # 212-2202
Supplier:   JONES FR
Test tubes, reductase
Catalog # 212-2202
Supplier:  JONES FR
Test tubes, reductase
Test tubes, reductase
Tubes with marks at 5 and 10 ml for Phosphatase and COD tests
Price
£180.00
Log in for availability
1 * 100 items
Product Details & Documents
Heavy wall rimless soda glass test tubes.
  • Yellow notation patch
  • Ceramic enamel ring marking at appropriate capacity
Specifications
  • Description:
    Tubes with marks at 5 and 10 ml for Phosphatase and COD tests
  • Seq:
    02