Test tubes, reductase
Tubes with marks at 2,5 and 10 ml for antibiotic tests
Catalog # 212-2201
Supplier: JONES FR
Jump to:
Product Details & Documents
Heavy wall rimless soda glass test tubes.
- Yellow notation patch
- Ceramic enamel ring marking at appropriate capacity
Specifications
- Description:Tubes with marks at 2,5 and 10 ml for antibiotic tests
- Seq:01