Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cycloheximide ≥90.0%

Cycloheximide ≥90.0%

Glass bottle, 10 g
Catalog # 441894C
Health
Toxic
Environment
Supplier:   VWR Chemicals
Cycloheximide ≥90.0%
Catalog # 441894C
Supplier:  VWR Chemicals
CAS Number:  66-81-9
Cycloheximide ≥90.0%
Cycloheximide ≥90.0%

Glass bottle, 10 g

CAS Number:  66-81-9
Price
£627.00
Log in for availability
1 * 10 g
Product Details & Documents

Inhibits DNA and protein synthesis.

Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I
Specifications

Specification Test Results

  • Assay:
    Min. 90.0 %
  • Melting point:
    107 - 120 °C

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.