Pyrotell multi test vial / 5ml lal per vial (0.06eu/ml) / approx 50 tests 1 * 1 items
CAPEG5006
Pyrotell multi test vial / 5ml lal per vial (0.06eu/ml) / approx 50 tests 1 * 1 items
CAPEG5006
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection