Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Transfer pipettes

Transfer pipette, not graduated, 6 ml, 225 mm, -
  613-4992
Transfer pipettes
  613-4992
Transfer pipettes
Transfer pipettes

Transfer pipette, not graduated, 6 ml, 225 mm, -

 
£64.00
   
1 * 400 items
 

PE, disposable.

  • Extra long or graduated
  • Sterile models are ethylene oxide sterilised
 
  • Sterile:
    -
  • Capacity:
    6 ml
  • Description:
    Transfer pipette, not graduated
  • Length:
    225 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.