Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Transfer pipettes

Transfer pipettes

Transfer pipette, not graduated, 23 ml, 300 mm, -
Catalog # 613-4987
Supplier:   MEDLINE SCIENTIFIC
Transfer pipettes
Catalog # 613-4987
Supplier:  MEDLINE SCIENTIFIC
Transfer pipettes
Transfer pipettes

Transfer pipette, not graduated, 23 ml, 300 mm, -

Price
£136.00
Log in for availability
1 * 100 items
Product Details & Documents

PE, disposable.

  • Extra long or graduated
  • Sterile models are ethylene oxide sterilised
Specifications
  • Sterile:
    -
  • Capacity:
    23 ml
  • Description:
    Transfer pipette, not graduated
  • Length:
    300 mm
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.