Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Transfer pipettes

Transfer pipettes

Transfer pipette, graduated (each ml), 5 ml, - mm, 216 mm, -
Catalog # 613-4977
Supplier:   MEDLINE SCIENTIFIC
Transfer pipettes
Catalog # 613-4977
Supplier:  MEDLINE SCIENTIFIC
Transfer pipettes
Transfer pipettes

Transfer pipette, graduated (each ml), 5 ml, - mm, 216 mm, -

Price
£51.30
Log in for availability
1 * 250 items
Product Details & Documents

PE, disposable.

  • Extra long or graduated
  • Sterile models are ethylene oxide sterilised
Specifications
  • Sterile:
    -
  • Capacity:
    5 ml
  • Description:
    Transfer pipette, graduated (each ml)
  • Length:
    216 mm
  • Seq:
    05
  • Stem Ø:
    - mm