Volumetric flasks, class B
1000 ml, 24/29, 0,8 ± ml
Catalog # 612-2248
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass, with plastic stopper.
Certifications: ISO 1042
Specifications
- NS:24/29
- Capacity:1000 ml
- Seq:07
- Tolerance:0,8 ± ml