Volumetric flasks, class B
500 ml, 19/26, 0,5 ± ml
Catalog # 612-2258
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass, with plastic stopper.
Certifications: ISO 1042
Specifications
- NS:19/26
- Capacity:500 ml
- Seq:06
- Tolerance:0,5 ± ml