Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

SPE columns, ISOLUTE® C2

C2, PP, 60 Å, 50 µm, 15 ml, 2000 mg
  ARGO320-0200-D
SPE columns, ISOLUTE® C2
  ARGO320-0200-D
SPE columns, ISOLUTE® C2

C2, PP, 60 Å, 50 µm, 15 ml, 2000 mg

 
 
   
1 * 20 items
 

ISOLUTE® C2 is a weakly non-polar sorbent for extraction of non-polar acidic, neutral and basic analytes from aqueous samples using a non-polar retention mechanism. It is used for the extraction of non-polar acidic, neutral and basic compounds from aqueous sample matrices. Secondary silanol interactions can be exploited to enhance extract purity and method robustness for basic analytes.

 
  • Particle Size:
    50 µm
  • Pore size:
    60 Å
  • Capacity:
    15 ml
  • Seq:
    11
  • Bed weight:
    2000 mg
  • Column hardware:
    PP
  • Phase:
    C2
Accessories

 

  •  
    Description
     
     
  • ARGO120-1103
    ISOLUTE® column adapter for 70 ml columns
       
    1 * 10 items - £98.90

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.