Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

SPE columns, ISOLUTE® C2

C2, PP, 60 Å, 50 µm, 6 ml, 1000 mg
  ARGO320-0100-C
SPE columns, ISOLUTE® C2
  ARGO320-0100-C
SPE columns, ISOLUTE® C2

C2, PP, 60 Å, 50 µm, 6 ml, 1000 mg

 
 
   
1 * 30 items
 

ISOLUTE® C2 is a weakly non-polar sorbent for extraction of non-polar acidic, neutral and basic analytes from aqueous samples using a non-polar retention mechanism. It is used for the extraction of non-polar acidic, neutral and basic compounds from aqueous sample matrices. Secondary silanol interactions can be exploited to enhance extract purity and method robustness for basic analytes.

 
  • Particle Size:
    50 µm
  • Pore size:
    60 Å
  • Capacity:
    6 ml
  • Seq:
    09
  • Bed weight:
    1000 mg
  • Column hardware:
    PP
  • Phase:
    C2
Accessories

 

  •  
    Description
     
     
  • ARGO120-1103
    ISOLUTE® column adapter for 70 ml columns
       
    1 * 10 items - £98.90