Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
SPE columns, ISOLUTE® C8(EC)

SPE columns, ISOLUTE® C8(EC)

C8(EC), PP, 60 Å, 50 µm, 70 ml, 100000 mg
Catalog # ARGO291-1000-F
SPE columns, ISOLUTE® C8(EC)
Catalog # ARGO291-1000-F
SPE columns, ISOLUTE® C8(EC)

C8(EC), PP, 60 Å, 50 µm, 70 ml, 100000 mg

Price
Inquire For Price
Log in for availability
1 * 16 items
Product Details & Documents

ISOLUTE® C8 endcapped is primarily used to extract molecules of wide ranging polarities from aqueous samples. When analytes are strongly retained by a C18 sorbent, the slightly weaker interaction with the C8(EC) can facilitate analyte elution with smaller elution volumes. It is a non-polar sorbent, manufactured using a silane and endcapped in order to reduce the polar secondary interactions associated with surface silanol groups.

Specifications
  • Particle Size:
    50 µm
  • Pore size:
    60 Å
  • Capacity:
    70 ml
  • Seq:
    20
  • Bed weight:
    100000 mg
  • Column hardware:
    PP
  • Phase:
    C8(EC)